Merck
  • Proteomics, Post-translational Modifications, and Integrative Analyses Reveal Molecular Heterogeneity within Medulloblastoma Subgroups.
Proteomics, Post-translational Modifications, and Integrative Analyses Reveal Molecular Heterogeneity within Medulloblastoma Subgroups.
Cancer cell (2018-09-12)
Tenley C Archer, Tobias Ehrenberger, Filip Mundt, Maxwell P Gold, Karsten Krug, Clarence K Mah, Elizabeth L Mahoney, Colin J Daniel, Alexander LeNail, Divya Ramamoorthy, Philipp Mertins, D R Mani, Hailei Zhang, Michael A Gillette, Karl Clauser, Michael Noble, Lauren C Tang, Jessica Pierre-François, Jacob Silterra, James Jensen, Pablo Tamayo, Andrey Korshunov, Stefan M Pfister, Marcel Kool, Paul A Northcott, Rosalie C Sears, Jonathan O Lipton, Steven A Carr, Jill P Mesirov, Scott L Pomeroy, Ernest Fraenkel
ABSTRACT
There is a pressing need to identify therapeutic targets in tumors with low mutation rates such as the malignant pediatric brain tumor medulloblastoma. To address this challenge, we quantitatively profiled global proteomes and phospho-proteomes of 45 medulloblastoma samples. Integrated analyses revealed that tumors with similar RNA expression vary extensively at the post-transcriptional and post-translational levels. We identified distinct pathways associated with two subsets of SHH tumors, and found post-translational modifications of MYC that are associated with poor outcomes in group 3 tumors. We found kinases associated with subtypes and showed that inhibiting PRKDC sensitizes MYC-driven cells to radiation. Our study shows that proteomics enables a more comprehensive, functional readout, providing a foundation for future therapeutic strategies.
Sigma-Aldrich
O-Phospho-L-tyrosine
Pricing and availability is not currently available.
Sigma-Aldrich
Nα-Acetyl-L-lysine
Pricing and availability is not currently available.