Merck
  • Immune Marker Profiling and Programmed Death Ligand 1 Expression Across NSCLC Mutations.
Immune Marker Profiling and Programmed Death Ligand 1 Expression Across NSCLC Mutations.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer (2018-09-30)
Maria I Toki, Nikita Mani, James W Smithy, Yuting Liu, Mehmet Altan, Brad Wasserman, Rasikh Tuktamyshov, Kurt Schalper, Konstantinos N Syrigos, David L Rimm
ABSTRACT
Programmed death 1/programmed death ligand 1 (PD-L1) axis inhibitors have been proven effective, especially in patients with tumors expressing PD-L1. Their clinical efficacy in patients with EGFR-activating mutations is still unclear, whereas KRAS mutations seem to be associated with good response. We used multiplexed quantitative immunofluorescence to investigate PD-L1 expression and to characterize tumor infiltrating lymphocyte (TIL) populations and their activation status in more than 150 NSCLC patients with known mutation status. PD-L1 expression was significantly lower in EGFR-mutant compared to KRAS-mutant, and EGFR/KRAS wild-type (WT) tumors. KRAS mutant tumors were more inflamed with higher CD4+, CD8+ and CD20+ TILs. Subgroup analysis by TIL activation status revealed that EGFR mutants had a high frequency of inactive TILs even though lymphocytes were present in the tumor microenvironment. In contrast, in KRAS mutants, when TILs were present they were almost always active. Additionally, we found differences between EGFR mutation sites in CD8+ expression and the TIL activation profile. Finally, activated EGFR correlated with increased PD-L1 expression in EGFR mutants but not in EGFR WT, whereas TIL activation was associated with higher PD-L1 only in EGFR/KRAS WT. Our findings show the unique immune profile of EGFR-mutant tumors. The high frequency of inactive TILs could explain the low immunotherapy response rates in these patients, whereas PD-L1 as a predictive biomarker may reflect the constitutive oncogenic signaling rather than immune signaling, which would be associated with high PD-L1 levels and TILs activation.
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection