Merck
  • A Neutralizing Antibody Recognizing Primarily N-Linked Glycan Targets the Silent Face of the HIV Envelope.
A Neutralizing Antibody Recognizing Primarily N-Linked Glycan Targets the Silent Face of the HIV Envelope.
Immunity (2018-03-20)
Tongqing Zhou, Anqi Zheng, Ulrich Baxa, Gwo-Yu Chuang, Ivelin S Georgiev, Rui Kong, Sijy O'Dell, Syed Shahzad-Ul-Hussan, Chen-Hsiang Shen, Yaroslav Tsybovsky, Robert T Bailer, Syna K Gift, Mark K Louder, Krisha McKee, Reda Rawi, Catherine H Stevenson, Guillaume B E Stewart-Jones, Justin D Taft, Eric Waltari, Yongping Yang, Baoshan Zhang, Sachin S Shivatare, Vidya S Shivatare, Chang-Chun D Lee, Chung-Yi Wu, James C Mullikin, Carole A Bewley, Dennis R Burton, Victoria R Polonis, Lawrence Shapiro, Chi-Huey Wong, John R Mascola, Peter D Kwong, Xueling Wu
ABSTRACT
Virtually the entire surface of the HIV-1-envelope trimer is recognized by neutralizing antibodies, except for a highly glycosylated region at the center of the "silent face" on the gp120 subunit. From an HIV-1-infected donor, #74, we identified antibody VRC-PG05, which neutralized 27% of HIV-1 strains. The crystal structure of the antigen-binding fragment of VRC-PG05 in complex with gp120 revealed an epitope comprised primarily of N-linked glycans from N262, N295, and N448 at the silent face center. Somatic hypermutation occurred preferentially at antibody residues that interacted with these glycans, suggesting somatic development of glycan recognition. Resistance to VRC-PG05 in donor #74 involved shifting of glycan-N448 to N446 or mutation of glycan-proximal residue E293. HIV-1 neutralization can thus be achieved at the silent face center by glycan-recognizing antibody; along with other known epitopes, the VRC-PG05 epitope completes coverage by neutralizing antibody of all major exposed regions of the prefusion closed trimer.
Sigma-Aldrich
1,6-Hexanediol, 99%
Supelco
Discovery® Glycan SPE Tube, bed wt. 250 mg, volume 3 mL, pk of 54
Supelco
Discovery® Glycan SPE Tube, bed wt. 1 g, volume 12 mL, pk of 20
Supelco
Discovery® Glycan SPE Tube, 5 g, volume 60 mL, pk of 16
Supelco
Discovery® Glycan SPE Bulk Packing, pkg of 50 g
Supelco
Discovery® Glycan SPE Tube, bed wt. 500 mg, volume 6 mL, pk of 30