Merck
  • Low-flow ischaemia and reperfusion in rat hearts: energetic of stunning and cardioprotection of genistein.
Low-flow ischaemia and reperfusion in rat hearts: energetic of stunning and cardioprotection of genistein.
The Journal of pharmacy and pharmacology (2018-06-23)
Germán A Colareda, Alicia E Consolini
ABSTRACT
Low-flow ischemia (LFI) is consequent to coronary disease and produces cardiac stunning during reperfusion (R). Energetic performance and mechanisms of Ca2+ handling during LFI/R are not known. Moreover, cardioprotection of the phytoestrogen genistein (Gen) remains to be demonstrated in LFI/R. The aim was to study the mechanisms of the stunning consequent to LFI/R and the effects of Gen on both sexes. Rat ventricles were perfused inside a calorimeter to measure maximal pressure development (P) and total heat rate (Ht) before and during exposition to LFI/R. The mechanisms of stunning were evaluated with selective drugs. Female hearts (FH) developed higher postischemic contractile recovery (PICR) and muscle economy (P/Ht) than males (MH). Cardioprotection was sensitive to blockade of mKATP channels, UCam and NOS. Perfusion of 20 μmol/l Gen reduced PICR and P/Ht during LFI/R in FH, and dysfunction was increased by mNCX blockade with mPTP opening. However, intraperitoneal 5 mg/kg Gen (Gen-ip) was cardioprotective in both sexes, and the beneficial effect of Gen-ip was blocked by 100 μmol/l 5-HD. FH are more protected than MH against the LFI/R dysfunction, which involves mitochondrial Ca2+ loss; Gen-ip was more cardioprotective in MH than in FH, mainly by activation of the mKATP channels.
Sigma-Aldrich
5-Hydroxydecanoic acid sodium salt, ≥97% (HPLC)
Pricing and availability is not currently available.