Merck
  • Bacterial exopolysaccharides as a modern biotechnological tool for modification of fungal laccase properties and metal ion binding.
Bacterial exopolysaccharides as a modern biotechnological tool for modification of fungal laccase properties and metal ion binding.
Bioprocess and biosystems engineering (2018-03-28)
Monika Osińska-Jaroszuk, Magdalena Jaszek, Magdalena Starosielec, Justyna Sulej, Anna Matuszewska, Monika Janczarek, Renata Bancerz, Jerzy Wydrych, Adrian Wiater, Anna Jarosz-Wilkołazka
ABSTRACT
Four bacterial EPSs extracted from Rhizobium leguminosarum bv. trifolii Rt24.2, Sinorhizobium meliloti Rm1021, Bradyrhizobium japonicum USDA110, and Bradyrhizobium elkanii USDA76 were determined towards their metal ion adsorption properties and possible modification of Cerrena unicolor laccase properties. The highest magnesium and iron ion-sorption capacity (~ 42 and ~ 14.5%, respectively) was observed for EPS isolated from B. japonicum USDA110. An evident influence of EPSs on the stability of laccase compared to the control values (without EPSs) was shown after 30-day incubation at 25 °C. The residual activity of laccases was obtained in the presence of Rh76EPS and Rh1021EPS, i.e., 49.5 and 41.5% of the initial catalytic activity, respectively. This result was confirmed by native PAGE electrophoresis. The EPS effect on laccase stability at different pH (from 3.8 to 7.0) was also estimated. The most significant changes at the optimum pH value (pH 5.8) was observed in samples of laccase stabilized by Rh76EPS and Rh1021EPS. Cyclic voltamperometry was used for analysis of electrochemical parameters of laccase stabilized by bacterial EPS and immobilized on single-walled carbon nanotubes (SWCNTs) with aryl residues. Laccases with Rh76EPS and Rh1021EPS had an evident shift of the value of the redox potential compared to the control without EPS addition. In conclusion, the results obtained in this work present a new potential use of bacterial EPSs as a metal-binding component and a modulator of laccase properties especially stability of enzyme activity, which can be a very effective tool in biotechnology and industrial applications.
Sigma-Aldrich
3,5-Dimethoxybenzaldehyde, 98%
Pricing and availability is not currently available.