Merck
  • Broadband (550-1350 nm) diffuse optical characterization of thyroid chromophores.
Broadband (550-1350 nm) diffuse optical characterization of thyroid chromophores.
Scientific reports (2018-07-04)
Sanathana Konugolu Venkata Sekar, Andrea Farina, Alberto Dalla Mora, Claus Lindner, Marco Pagliazzi, Mireia Mora, Gloria Aranda, Hamid Dehghani, Turgut Durduran, Paola Taroni, Antonio Pifferi
ABSTRACT
Thyroid plays an important role in the endocrine system of the human body. Its characterization by diffuse optics can open new path ways in the non-invasive diagnosis of thyroid pathologies. Yet, the absorption spectra of tyrosine and thyroglobulin-key tissue constituents specific to the thyroid organ-in the visible to near infrared range are not fully available. Here, we present the optical characterization of tyrosine (powder), thyroglobulin (granular form) and iodine (aqueous solution) using a time domain broadband diffuse optical spectrometer in the 550-1350 nm range. Various systematic errors caused by physics of photo migration and sample inherent properties were effectively suppressed by means of advanced time domain diffuse optical methods. A brief comparison with various other known tissue constituents is presented, which reveals key spectral regions for the quantification of the thyroid absorbers in an in vivo scenario.
Sigma-Aldrich
Thyroglobulin from porcine thyroid gland, powder
Pricing and availability is not currently available.