Merck
  • Visit-to-visit cholesterol variability correlates with coronary atheroma progression and clinical outcomes.
Visit-to-visit cholesterol variability correlates with coronary atheroma progression and clinical outcomes.
European heart journal (2018-04-25)
Donald Clark, Stephen J Nicholls, Julie St John, Mohamed B Elshazly, Samir R Kapadia, E Murat Tuzcu, Steven E Nissen, Rishi Puri
ABSTRACT
Utilizing serial intravascular ultrasonography (IVUS), we aimed to exam the association of intra-individual lipid variability, coronary atheroma progression, and clinical outcomes. We performed a post hoc patient-level analysis of nine clinical trials involving 4976 patients with coronary artery disease who underwent serial coronary IVUS in the setting of a range of medical therapies. We assessed the associations between progression in percent atheroma volume (ΔPAV), clinical outcomes, and visit-to-visit lipid variability including low-density lipoprotein cholesterol (LDL-C), high-density lipoprotein cholesterol (HDL-C), non-HDL-C, total cholesterol (TC)/HDL-C, and apolipoprotein B (ApoB). Variability of lipid parameters was measured using intra-individual standard deviation over 3, 6, 12, 18, and 24 months. Atherogenic lipoprotein variability significantly associated with ΔPAV [odds ratio (95% confidence interval; P-value), LDL-C: 1.09 (1.02, 1.17, P = 0.01); non-HDL-C: 1.10 (1.02, 1.18, P = 0.01); TC/HDL-C: 1.14 (1.06, 1.24, P = 0.001); ApoB: 1.13 (1.03, 1.24, P = 0.01)]. Survival curves revealed significant stepwise relationships between cumulative major adverse cardiovascular events and increasing quartiles of atherogenic lipoprotein variability at 24-months follow-up (log-rank P < 0.01 for all lipoproteins except HDL-C). Stronger associations were noted between achieved lipoprotein levels and ΔPAV [LDL-C: 1.27 (1.17, 1.39; P < 0.001); non-HDL-C: 1.32 (1.21, 1.45; P < 0.001); TC/HDL-C: 1.31 (1.19, 1.45; P < 0.001); ApoB: 1.20 (1.07, 1.35; P = 0.003)]. Greater visit-to-visit variability in atherogenic lipoprotein levels significantly associates with coronary atheroma progression and clinical outcomes, although the association between achieved atherogenic lipoproteins and atheroma progression appears stronger. These data highlight the importance of achieving low and consistent atherogenic lipoprotein levels to promote plaque regression and improve clinical outcomes.