Merck
  • Cyclin E2-CDK2 mediates SAMHD1 phosphorylation to abrogate its restriction of HBV replication in hepatoma cells.
Cyclin E2-CDK2 mediates SAMHD1 phosphorylation to abrogate its restriction of HBV replication in hepatoma cells.
FEBS letters (2018-05-22)
Jie Hu, Miao Qiao, Yanmeng Chen, Hua Tang, Wenlu Zhang, Dan Tang, Sidie Pi, Juan Dai, Ni Tang, Ailong Huang, Yuan Hu
ABSTRACT
SAMHD1 inhibits Hepatitis B virus (HBV) replication by reducing the intracellular dNTP levels. However, how SAMHD1 phosphorylation is regulated to abrogate its restriction of HBV replication in hepatoma cells is poorly understood. Here, we show that HBV replication and SAMHD1 phosphorylation levels are significantly reduced by knocking down cyclin-dependent kinase (CDK) 2 expression or in the presence of a CDK2 inhibitor. SAMHD1 binds to CDK2 in hepatocarcinoma cells, and this interaction does not require the HBV core protein. Furthermore, cyclin E2 participates in regulating viral replication through the CDK2/SAMHD1 phosphorylation pathway in an HBV infection system. Collectively, our results provide evidence that CDK2 has a greater role in regulating SAMHD1 phosphorylation and HBV replication than CDK1 or CDK6.
Product Description
Millipore
Protein G Agarose, Fast Flow, Protein G Agarose, Fast Flow suitable for medium and low pressure chromatography of IgG from mouse, sheep, and rabbit, and for immunoprecipitations.