Merck
  • Alternative assembly of respiratory complex II connects energy stress to metabolic checkpoints.
Alternative assembly of respiratory complex II connects energy stress to metabolic checkpoints.
Nature communications (2018-06-09)
Ayenachew Bezawork-Geleta, He Wen, LanFeng Dong, Bing Yan, Jelena Vider, Stepana Boukalova, Linda Krobova, Katerina Vanova, Renata Zobalova, Margarita Sobol, Pavel Hozak, Silvia Magalhaes Novais, Veronika Caisova, Pavel Abaffy, Ravindra Naraine, Ying Pang, Thiri Zaw, Ping Zhang, Radek Sindelka, Mikael Kubista, Steven Zuryn, Mark P Molloy, Michael V Berridge, Karel Pacak, Jakub Rohlena, Sunghyouk Park, Jiri Neuzil
ABSTRACT
Cell growth and survival depend on a delicate balance between energy production and synthesis of metabolites. Here, we provide evidence that an alternative mitochondrial complex II (CII) assembly, designated as CIIlow, serves as a checkpoint for metabolite biosynthesis under bioenergetic stress, with cells suppressing their energy utilization by modulating DNA synthesis and cell cycle progression. Depletion of CIIlow leads to an imbalance in energy utilization and metabolite synthesis, as evidenced by recovery of the de novo pyrimidine pathway and unlocking cell cycle arrest from the S-phase. In vitro experiments are further corroborated by analysis of paraganglioma tissues from patients with sporadic, SDHA and SDHB mutations. These findings suggest that CIIlow is a core complex inside mitochondria that provides homeostatic control of cellular metabolism depending on the availability of energy.
Sigma-Aldrich
Anti-ATP5B antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SDHAF4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder