Merck
  • AURKA Suppresses Leukemic THP-1 Cell Differentiation through Inhibition of the KDM6B Pathway.
AURKA Suppresses Leukemic THP-1 Cell Differentiation through Inhibition of the KDM6B Pathway.
Molecules and cells (2018-02-27)
Jin Woo Park, Hana Cho, Hyein Oh, Ji-Young Kim, Sang-Beom Seo
ABSTRACT
Aberrations in histone modifications are being studied in mixed-lineage leukemia (MLL)-AF9-driven acute myeloid leukemia (AML). In this study, we focused on the regulation of the differentiation of the MLL-AF9 type AML cell line THP-1. We observed that, upon phorbol 12-myristate 13-acetate (PMA) treatment, THP-1 cells differentiated into monocytes by down-regulating Aurora kinase A (AURKA), resulting in a reduction in H3S10 phosphorylation. We revealed that the AURKA inhibitor alisertib accelerates the expression of the H3K27 demethylase KDM6B, thereby dissociating AURKA and YY1 from the KDM6B promoter region. Using Flow cytometry, we found that alisertib induces THP-1 differentiation into monocytes. Furthermore, we found that treatment with the KDM6B inhibitor GSK-J4 perturbed the PMA-mediated differentiation of THP-1 cells. Thus, we discovered the mechanism of AURKA-KDM6B signaling that controls the differentiation of THP-1 cells, which has implications for biotherapy for leukemia.
Sigma-Aldrich
GSK-J4, ≥98% (HPLC)
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys36) Antibody, serum, Upstate®