Merck
  • Properties of PC4 and an RNA polymerase II complex in directing activated and basal transcription in vitro.
Properties of PC4 and an RNA polymerase II complex in directing activated and basal transcription in vitro.
The Journal of biological chemistry (1998-06-20)
S Y Wu, C M Chiang
ABSTRACT
A human RNA polymerase II (pol II) complex was isolated from a HeLa-derived cell line that conditionally expresses an epitope-tagged RPB9 subunit of human pol II. The isolated FLAG-tagged pol II complex (f:pol II) contains a subset of general transcription factors but is devoid of TFIID and TFIIA. In conjunction with TATA-binding protein (TBP) or TFIID, f:pol II is able to mediate both basal and activated transcription by Gal4-VP16 when a transcriptional coactivator PC4 is also provided. Interestingly, PC4, in the absence of a transcriptional activator, actually functions as a repressor to inhibit basal transcription. Remarkably, TBP is able to mediate activator function in this transcription system. The presence of TBP-associated factors, however, helps overcome PC4 repression and further enhance the level of activation mediated by TBP. Alleviation of PC4 repression can also be achieved by preincubation of the transcriptional components with the DNA template. Sarkosyl disruption of preinitiation complex formation further illustrates that PC4 can only inhibit transcription prior to the assembly of a functional preinitiation complex. These results suggest that PC4 represses basal transcription by preventing the assembly of a functional preinitiation complex, but it has no effect on the later steps of the transcriptional process.
Sigma-Aldrich
PC4, F77P mutant human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Pricing and availability is not currently available.