Merck
  • BRG1 contains a conserved domain of the SWI2/SNF2 family necessary for normal mitotic growth and transcription.
BRG1 contains a conserved domain of the SWI2/SNF2 family necessary for normal mitotic growth and transcription.
Nature (1993-11-11)
P A Khavari, C L Peterson, J W Tamkun, D B Mendel, G R Crabtree
ABSTRACT
Sequence-specific DNA binding activators of gene transcription may be assisted by SWI2 (SNF2), which contains a DNA-dependent ATPase domain. We have isolated a human complementary DNA encoding a 205K nuclear protein, BRG1, that contains extensive homology to SWI2 and Drosophila brahma. We report here that a SWI2/BRG1 chimera with the DNA-dependent ATPase domain replaced by corresponding human sequence restored normal mitotic growth and capacity for transcriptional activation to swi2- yeast cells. Point mutation of the conserved ATP binding site lysine abolished this complementation. This mutation in SWI2 exerted a dominant negative effect on transcription in yeast. A lysine to arginine substitution at the corresponding residue of BRG1 also generated a transcriptional dominant negative in human cells. BRG1 is exclusively nuclear and present in a high M(r) complex of about 2 x 10(6). These results show that the SWI2 family DNA-dependent ATPase domain has functional conservation between yeast and humans and suggest that a SWI/SNF protein complex is required for the activation of selective mammalian genes.
Sigma-Aldrich
BRG1, wild type human, recombinant, expressed in insect cells, ≥80% (SDS-PAGE)