Merck
  • PIM2 interacts with tristetraprolin and promotes breast cancer tumorigenesis.
PIM2 interacts with tristetraprolin and promotes breast cancer tumorigenesis.
Molecular oncology (2018-03-24)
Chune Ren, Tingting Yang, Pengyun Qiao, Li Wang, Xue Han, Shijun Lv, Yonghong Sun, Zhijun Liu, Yu Du, Zhenhai Yu
ABSTRACT
Tristetraprolin (TTP) is an AU-rich element-binding protein that regulates mRNA stability and plays important roles in cancer. The mechanisms by which TTP is regulated in breast cancer are poorly understood. Using multiple biochemical approaches, we found that proviral insertion in murine lymphomas 2 (PIM2) is a novel binding partner of TTP. Interestingly, PIM2 decreased TTP protein levels independent of its kinase activity. PIM2 instead targeted TTP protein for degradation via the ubiquitin-proteasome pathway. Furthermore, immunohistochemical staining showed that PIM2 and TTP protein levels were negatively correlated in human breast cancer samples. Indeed, PIM2 overexpression de-repressed TTP-mediated inhibition of breast cancer cell proliferation and migration in vitro and promoted breast tumor xenograft growth in vivo. These findings demonstrate an important role for the PIM2-TTP complex in breast cancer tumorigenesis, suggesting that PIM2 may represent a potential therapeutic target for breast cancer treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human PIM2
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%