Merck
  • Segmentation of the zebrafish axial skeleton relies on notochord sheath cells and not on the segmentation clock.
Segmentation of the zebrafish axial skeleton relies on notochord sheath cells and not on the segmentation clock.
eLife (2018-04-07)
Laura Lleras Forero, Rachna Narayanan, Leonie Fa Huitema, Maaike VanBergen, Alexander Apschner, Josi Peterson-Maduro, Ive Logister, Guillaume Valentin, Luis G Morelli, Andrew C Oates, Stefan Schulte-Merker
ABSTRACT
Segmentation of the axial skeleton in amniotes depends on the segmentation clock, which patterns the paraxial mesoderm and the sclerotome. While the segmentation clock clearly operates in teleosts, the role of the sclerotome in establishing the axial skeleton is unclear. We severely disrupt zebrafish paraxial segmentation, yet observe a largely normal segmentation process of the chordacentra. We demonstrate that axial entpd5+ notochord sheath cells are responsible for chordacentrum mineralization, and serve as a marker for axial segmentation. While autonomous within the notochord sheath, entpd5 expression and centrum formation show some plasticity and can respond to myotome pattern. These observations reveal for the first time the dynamics of notochord segmentation in a teleost, and are consistent with an autonomous patterning mechanism that is influenced, but not determined by adjacent paraxial mesoderm. This behavior is not consistent with a clock-type mechanism in the notochord.
Sigma-Aldrich
Ethyl 3-aminobenzoate, 97%