Merck
  • Increased Microglial CSF1R Expression in the SIV/Macaque Model of HIV CNS Disease.
Increased Microglial CSF1R Expression in the SIV/Macaque Model of HIV CNS Disease.
Journal of neuropathology and experimental neurology (2018-01-11)
Audrey C Knight, Samuel A Brill, Suzanne E Queen, Patrick M Tarwater, Joseph L Mankowski
ABSTRACT
Chronic microglial activation and associated neuroinflammation are key factors in neurodegenerative diseases including HIV-associated neurocognitive disorders. Colony stimulating factor 1 receptor (CSF1R)-mediated signaling is constitutive in cells of the myeloid lineage, including microglia, promoting cell survival, proliferation, and differentiation. In amyotrophic lateral sclerosis and Alzheimers disease, CSF1R is upregulated. Inhibiting CSF1R signaling in animal models of these diseases improved disease outcomes. In our studies, CNS expression of the CSF1R ligand, colony-stimulating factor 1 (CSF1) was significantly increased in a SIV/macaque model of HIV CNS disease. Using a Nanostring nCounter immune panel, we found CSF1 overexpression was strongly correlated with upregulation of microglial genes involved in antiviral and oxidative stress responses. Using in situ hybridization, we found that CSF1R mRNA was only present in Iba-1 positive microglia. By ELISA and immunostaining with digital image analysis, SIV-infected macaques had significantly higher CSF1R levels in frontal cortex than uninfected macaques (p = 0.018 and p = 0.02, respectively). SIV-infected macaques treated with suppressive ART also had persistently elevated CSF1R similar to untreated SIV-infected macaques. Coordinate upregulation of CSF1 and CSF1R expression implicates this signaling pathway in progressive HIV CNS disease.
Sigma-Aldrich
Fumitremorgin C, from Neosartorya fischeri, film
Sigma-Aldrich
Human CSF1R / Macrophage Colony-stimulating Factor 1 Receptor ELISA Kit