Merck
  • Fate of leaf litter deposits and impacts on oxygen availability in bank filtration column studies.
Fate of leaf litter deposits and impacts on oxygen availability in bank filtration column studies.
Environmental research (2018-03-31)
Uranchimeg Bayarsaikhan, Josefine Filter, Ulrich Gernert, Martin Jekel, Aki Sebastian Ruhl
ABSTRACT
Degradation of particulate organic carbon (POC) such as leaf litter might deplete dissolved oxygen within the upper layers of bank filtration, an efficient and robust barrier for pathogens and for various organic micro-pollutants (OMP) in water supply systems worldwide. The degradation of OMP during bank filtration depends on the redox conditions. The present study aimed at identifying the impacts and fates of different local leaves on the oxygen consumption and the possible biological degradation of indicator OMP. Oxygen concentrations initially decreased within the columns from around 8 mg/L in the influent to low concentrations indicating extensive consumption within a short travel distance. Still a substantial oxygen consumption was observed after 250 days. OMP concentrations were not significantly affected by the microbial processes. A layer of calcium carbonate crystallites was observed on the POC layer. Some leaf fragments appeared to be persistant towards degradation and the carbon content relative to nitrogen and sulfur contents decreased within 250 days. The results demonstrate that trees at bank filtration sites might have a strong long-term impact on the subsurface redox conditions.
Sigma-Aldrich
Orotidine 5′-monophosphate trisodium salt, ≥99% (HPLC), powder