Merck
  • Impact of Phospholipids and Tocopherols on the Oxidative Stability of Soybean Oil-in-Water Emulsions.
Impact of Phospholipids and Tocopherols on the Oxidative Stability of Soybean Oil-in-Water Emulsions.
Journal of agricultural and food chemistry (2018-04-10)
Gautam K Samdani, D Julian McClements, Eric A Decker
ABSTRACT
Phospholipids have been shown to act synergistically with tocopherols and delay lipid oxidation in bulk oil. The synergistic activity between phospholipids and tocopherols is due to the ability of amino-group-containing phospholipids (e.g., phosphatidylethanolamine (PE) and phosphatidylserine (PS)) to convert oxidized tocopherol back into tocopherols. This study shows the effect of PE and PS on the antioxidant activity of different tocopherol homologues in oil-in-water emulsions. Effect of emulsifier type on the interaction between tocopherols and phospholipids was also studied. δ-Tocopherol and PE exhibited greater antioxidant activity as compared to α-tocopherol and PE. PS displayed 1.5-3 times greater synergism than PE with Tween 20 as emulsifier whereas both PE and PS had a similar antioxidant activity in the presence of α-tocopherol when bovine serum albumin was used as the emulsifier. This study is the first to show that PE and PS can act synergistically with tocopherols to inhibit lipid oxidation in oil-in-water emulsions and can present a new clean label antioxidant strategy for food emulsions.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand