Merck
  • Assessment of the pharmacological interactions between the nematodicidal fenbendazole and the flukicidal triclabendazole: In vitro studies with bovine liver microsomes and slices.
Assessment of the pharmacological interactions between the nematodicidal fenbendazole and the flukicidal triclabendazole: In vitro studies with bovine liver microsomes and slices.
Journal of veterinary pharmacology and therapeutics (2018-02-22)
P Viviani, A L Lifschitz, M L Maté, J P García, C E Lanusse, G L Virkel
ABSTRACT
Parasitic diseases have a significant impact on livestock production. Nematodicidal drugs, such as fenbendazole (FBZ) or its oxidized metabolite oxfendazole (OFZ), can be used along with the trematodicidal triclabendazole (TCBZ), to broaden the spectrum of anthelmintic activity. However, co-exposure to these compounds could lead to drug-drug (D-D) interactions and eventually alter the clinical profile of each active principle. The aim of this study was to assess the presence of such interactions by means of two in vitro models, namely bovine liver microsomal fractions and bovine precision-cut liver slices (PCLSs). To this end, an in vitro assessment involving incubation of FBZ and TCBZ or a combination of FBZ and TCBZ was carried out. Results with microsomal fractions showed a 78.4% reduction (p = .002) in the rate of OFZ production upon co-incubation, whereas the sulfoxide metabolite of TCBZ (TCBZSO) exhibited a decreasing tendency. With PCLS, OFZ accumulation in the incubation medium increased 1.8-fold upon co-incubation, whereas TCBZSO accumulation decreased by 28%. The accumulation of FBZ and OFZ in the liver tissue increased upon 2-hr co-incubation, from 2.1 ± 1.5 to 18.2 ± 6.1 (p = .0009) and from 0.4 ± 0.1 to 1.3 ± 0.3 nmol (p = .0005), respectively. These results confirm the presence of D-D interactions between FBZ and TCBZ. Further studies are needed to determine the extent of involvement of drug-metabolizing enzymes and membrane transporters in interactions between compounds largely used in livestock production systems.
Product Description
Supelco
Triclabendazole sulfoxide, VETRANAL®, analytical standard
Supelco
Triclabendazole sulfone, VETRANAL®, analytical standard
Supelco
Fenbendazole sulfone, VETRANAL®, analytical standard