Merck
  • Assay of the set of all Sudan azodye (I, II, III, IV, and Para-Red) contaminating agents by liquid chromatography-tandem mass spectrometry and isotope dilution methodology.
Assay of the set of all Sudan azodye (I, II, III, IV, and Para-Red) contaminating agents by liquid chromatography-tandem mass spectrometry and isotope dilution methodology.
Journal of agricultural and food chemistry (2007-12-07)
Fabio Mazzotti, Leonardo Di Donna, Loredana Maiuolo, Anna Napoli, Raffaele Salerno, Ashiff Sajjad, Giovanni Sindona
ABSTRACT
A high-throughput mass spectrometric method is presented for the simultaneous detection of Sudan I, II, III, IV, and Para-Red azodyes in foodstuff. The method is based on the use of deuterium-labeled internal standards and atmospheric pressure chemical ionization (APCI) in a triple-quadrupole instrument. The gas-phase breakdown pattern of each labeled and unlabeled analogue displays the naphthoic moiety as a common fragment. The search for the parents of this common species (parent ion scans) allows, by flow injection and in a single run, the evaluation of the presence of each polluting species spiked in typical foodstuffs. A detailed assay of each azodye was performed by LC-APCI and isotope dilution method, through the multiple reaction monitoring approach, using deuterium-labeled internal standards. Sudan dyes can be quantified above the threshold of 10 ppb except for Sudan Para-Red, for which the limit of quantification was 20 ppb, likely due to the different ionization efficiency.
Sigma-Aldrich
Sudan IV, BioXtra, certified by the BSC
Sigma-Aldrich
Sudan IV, Certified Biological Stain Commission,Dye content ≥80 %
Supelco
Sudan IV, analytical standard
Sigma-Aldrich
Para Red, Dye content 95 %