Merck
  • Effects of pinostrobin on estrogen metabolism and estrogen receptor transactivation.
Effects of pinostrobin on estrogen metabolism and estrogen receptor transactivation.
Cancer letters (2000-06-07)
J C Le Bail, L Aubourg, G Habrioux
ABSTRACT
The interaction between the estrogen receptor and 5-hydroxy-7-methoxyflavanone (pinostrobin) was studied in the presence or absence of estradiol or dehydroepiandrosterone sulfate (DHEAS), respectively, using a stably transfected human breast cancer cell line (MVLN). We also evaluated its action on the proliferation in estrogen-dependent (MCF-7) human breast cancer cells in the same conditions than the estrogen receptor assay. On the other hand pinostrobin was evaluated for their effects on the human placental aromatase, 3beta-hydroxysteroid dehydrogenase Delta(4)/Delta(5) isomerase and 17beta-hydroxysteroid dehydrogenase activities. Pinostrobin did not possess antiestrogenic activity but presented anti-aromatase activity and decreased the growth of MCF-7 cells induced by DHEAS and E(2). This study provides particularly evidence of the potential biological interest of pinostrobin among the flavonoids.