- Staining reaction for beta-galactosidase in immunocytochemistry and in situ hybridization.
Staining reaction for beta-galactosidase in immunocytochemistry and in situ hybridization.
European journal of histochemistry : EJH (1992-01-01)
P Gugliotta, D Pacchioni, G Bussolati
PMID1380848
ABSTRACT
beta-galactosidase, revealed by an indigo blue reaction product, represents a valid tracer in immunohistochemistry. Observations on the instability of the indigo precipitate led us to investigate this phenomenon. We conclude that avoiding of xylene, and mounting of the preparates in Histoclear (a xylene substitute) and Canada balsam (instead of synthetic resin mountants) yields a sharp and stable indigo precipitate. In addition, we propose a sensitive alternative staining procedure for beta-galactosidase, based on TNBT reduction and precipitation. These reactions can be used both in immunohistochemistry and in in situ hybridization.
MATERIALS