Merck
  • Elevated Adenylyl Cyclase 9 Expression Is a Potential Prognostic Biomarker for Patients with Colon Cancer.
Elevated Adenylyl Cyclase 9 Expression Is a Potential Prognostic Biomarker for Patients with Colon Cancer.
Medical science monitor : international medical journal of experimental and clinical research (2018-01-03)
Hua Yi, Kun Wang, Jun-Feng Jin, He Jin, Lihua Yang, Yidan Zou, Biaoyan Du, Xiaodong Liu
ABSTRACT
BACKGROUND Adenylyl cyclase 9 (ADCY9) is an enzyme that modulates signal transduction by producing the second messenger, cyclic adenosine monophosphate (cAMP). The aim of the present study was to investigate the association of ADCY9 expression with clinicopathological features and disease-free survival of colon cancer patients. MATERIAL AND METHODS Immunohistochemistry staining with ADCY9 antibody was performed on a tissue microarray. Immunoreactivity scores (IRS) were recorded and applied for association analysis. ADCY9 mRNA expression and clinicopathogical information were also extracted from TCGA colon cancer dataset and analyzed using univariate and multivariate Cox proportional hazards models.  RESULTS ADCY9 IRS was significantly higher (P=0.002) in tumor tissues (6.40±1.26, n=200) than in adjacent normal samples (4.13±0.83, n=8). The IRS and mRNA expression of ADCY9 were correlated to colon cancer TNM staging. Longer disease-free survival was observed in patients with lower ADCY9 expression (P=0.001). In the multivariate models, ADCY9 expression level (hazard ratio [HR] 5.495, 95% confidence interval [CI] 1.753-17.227, P=0.003), and distant metastasis (HR 4.329, 95% CI 1.374-13.636, P=0.012) were still associated with disease-free survival. CONCLUSIONS High ADCY9 expression is a poor prognostic factor for disease-free survival in colon cancer.
Sigma-Aldrich
Anti-ADCY9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution