Merck
  • Effect of Ethephon, Indole Butyric Acid, and Treatment Solution pH on Rooting and on Ethylene Levels within Mung Bean Cuttings.
Effect of Ethephon, Indole Butyric Acid, and Treatment Solution pH on Rooting and on Ethylene Levels within Mung Bean Cuttings.
Plant physiology (1978-02-01)
K W Mudge, B T Swanson
ABSTRACT
Light-grown mung bean (Phaseolus aureus Roxb.) cuttings were treated with buffered and nonbuffered solutions of Ethephon, indole butyric acid (IBA), and the combination of both. Ethephon treatment resulted in increased tissue ethylene levels with increasing solution pH, but had no effect on rooting. IBA treatment had no effect on tissue ethylene levels, but strongly promoted rooting. Combinations of Ethephon and IBA had no effect on rooting of mung bean cuttings beyond that obtained by IBA alone.
Sigma-Aldrich
Indole-3-butyric acid, ≥98.0% (T)