Merck
  • A short Gfi-1B isoform controls erythroid differentiation by recruiting the LSD1-CoREST complex through the dimethylation of its SNAG domain.
A short Gfi-1B isoform controls erythroid differentiation by recruiting the LSD1-CoREST complex through the dimethylation of its SNAG domain.
Journal of cell science (2012-03-09)
Benoît Laurent, Voahangy Randrianarison-Huetz, Emilie Frisan, Charlotte Andrieu-Soler, Eric Soler, Michaela Fontenay, Isabelle Dusanter-Fourt, Dominique Duménil
ABSTRACT
Gfi-1B is a transcriptional repressor essential for the regulation of erythropoiesis and megakaryopoiesis. Here we identify Gfi-1B p32, a Gfi-1B isoform, as essential for erythroid differentiation. Gfi-1B p32 is generated by alternative splicing and lacks the two first zinc finger domains of the protein. Selective knock down of Gfi-1B p32 compromises erythroid differentiation, whereas its ectopic expression induces erythropoiesis in the absence of erythropoietin. Gfi-1B p32 isoform binds to Gfi-1B target gene promoters and associates with the LSD1-CoREST repressor complex more efficiently than the major Gfi-1B p37 isoform. Furthermore, we show that Gfi-1B includes a KSKK motif in its SNAG domain, which recruits the repressor complex only when dimethylated on lysine 8. Mutation of lysine 8 prevents Gfi-1B p32-induced erythroid development. Our results thus highlight a key role for the alternatively spliced Gfi-1B p32 isoform in erythroid development.
Sigma-Aldrich
Anti-CoREST Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Pricing and availability is not currently available.