Merck
  • Extensive degradation of myelin basic protein isoforms by calpain following traumatic brain injury.
Extensive degradation of myelin basic protein isoforms by calpain following traumatic brain injury.
Journal of neurochemistry (2006-08-09)
Ming Cheng Liu, Veronica Akle, Wenrong Zheng, Jason Kitlen, Barbara O'Steen, Stephen F Larner, Jitendra R Dave, Frank C Tortella, Ronald L Hayes, Kevin K W Wang
ABSTRACT
Axonal injury is one of the key features of traumatic brain injury (TBI), yet little is known about the integrity of the myelin sheath. We report that the 21.5 and 18.5-kDa myelin basic protein (MBP) isoforms degrade into N-terminal fragments (of 10 and 8 kDa) in the ipsilateral hippocampus and cortex between 2 h and 3 days after controlled cortical impact (in a rat model of TBI), but exhibit no degradation contralaterally. Using N-terminal microsequencing and mass spectrometry, we identified a novel in vivo MBP cleavage site between Phe114 and Lys115. A MBP C-terminal fragment-specific antibody was then raised and shown to specifically detect MBP fragments in affected brain regions following TBI. In vitro naive brain lysate and purified MBP digestion showed that MBP is sensitive to calpain, producing the characteristic MBP fragments observed in TBI. We hypothesize that TBI-mediated axonal injury causes secondary structural damage to the adjacent myelin membrane, instigating MBP degradation. This could initiate myelin sheath instability and demyelination, which might further promote axonal vulnerability.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 119-131, clone 2, culture supernatant, clone 2, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-MMP-2 Antibody, a.a. 468-483 hMMP2, clone 42-5D11, clone 42-5D11, Chemicon®, from mouse
Pricing and availability is not currently available.