Merck
  • Electrochemical Nickel Catalysis for Sp2-Sp3 Cross-Electrophile Coupling Reactions of Unactivated Alkyl Halides.
Electrochemical Nickel Catalysis for Sp2-Sp3 Cross-Electrophile Coupling Reactions of Unactivated Alkyl Halides.
Organic letters (2017-07-14)
Robert J Perkins, Dylan J Pedro, Eric C Hansen
ABSTRACT
A constant-current electrochemical method for reducing catalytic nickel complexes in sp2-sp3 cross-electrophile coupling reactions has been developed. The electrochemical reduction provides reliable nickel catalyst activation and turnover and offers a tunable parameter for reaction optimization, in contrast to more standard activated metal powder reductants. The electrochemical reactions give yields (i.e., 51-86%) and selectivities as high or superior to those using metal powder reductants and provide access to a wider substrate scope.
Sigma-Aldrich
Pyridine-2,6-bis(carboximidamide) dihydrochloride, ≥95%
Sigma-Aldrich
4-Methoxypicolinimidamide hydrochloride, ≥95%