Merck
  • Host defense peptide LL-37 selectively reduces proinflammatory macrophage responses.
Host defense peptide LL-37 selectively reduces proinflammatory macrophage responses.
Journal of immunology (Baltimore, Md. : 1950) (2011-03-29)
Kelly L Brown, Grace F T Poon, Darlene Birkenhead, Olga M Pena, Reza Falsafi, Claes Dahlgren, Anna Karlsson, Johan Bylund, Robert E W Hancock, Pauline Johnson
ABSTRACT
The human cathelicidin peptide, LL-37, is a host defense peptide with a wide range of immunomodulatory activities and modest direct antimicrobial properties. LL-37 can exert both pro- and anti-inflammatory effects and can modulate the proinflammatory responses of human peripheral blood monocytes and epithelial cells. In this study, we evaluated the effect of LL-37 on mouse bone marrow-derived macrophages (BMDM) and tissue macrophages in vitro and in vivo. LL-37 dramatically reduced TNF-α and NO levels produced by LPS and IFN-γ-polarized M1-BMDM and slightly reduced reactive oxygen species production by these cells. LL-37 did not affect the ability of IL-4-polarized M2-BMDM to upregulate arginase activity, although it did inhibit LPS-induced TNF-α secretion in these cells. LL-37 did not compromise the ability of M1-polarized BMDM to phagocytose and kill bacteria and did not affect the uptake of apoptotic neutrophils by M2-polarized BMDM. However, LL-37-treated M1-BMDM were more efficient at suppressing tumor growth in vitro. LL-37 significantly reduced LPS-induced TNF-α secretion in ex vivo alveolar macrophages, whereas its effect on peritoneal macrophages was much less dramatic. Effective inhibition of LPS-induced TNF-α secretion by alveolar macrophages also occurred in vivo when LL-37 was administered by intratracheal injection. This demonstrates a selective ability of LL-37 to decrease M1-BMDM, M2-BMDM, and tissue macrophage production of the proinflammatory cytokine TNF-α in response to LPS while leaving other crucial anti-inflammatory M1 and M2 macrophage functions unaltered.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Pricing and availability is not currently available.