Merck
  • Structural basis for selectivity and diversity in angiotensin II receptors.
Structural basis for selectivity and diversity in angiotensin II receptors.
Nature (2017-04-06)
Haitao Zhang, Gye Won Han, Alexander Batyuk, Andrii Ishchenko, Kate L White, Nilkanth Patel, Anastasiia Sadybekov, Beata Zamlynny, Michael T Rudd, Kaspar Hollenstein, Alexandra Tolstikova, Thomas A White, Mark S Hunter, Uwe Weierstall, Wei Liu, Kerim Babaoglu, Eric L Moore, Ryan D Katz, Jennifer M Shipman, Margarita Garcia-Calvo, Sujata Sharma, Payal Sheth, Stephen M Soisson, Raymond C Stevens, Vsevolod Katritch, Vadim Cherezov
ABSTRACT
The angiotensin II receptors AT
Sigma-Aldrich
ZD7155 hydrochloride, ≥97% (HPLC)