Merck
  • Differential activation and antagonistic function of HIF-{alpha} isoforms in macrophages are essential for NO homeostasis.
Differential activation and antagonistic function of HIF-{alpha} isoforms in macrophages are essential for NO homeostasis.
Genes & development (2010-03-03)
Norihiko Takeda, Ellen L O'Dea, Andrew Doedens, Jung-whan Kim, Alexander Weidemann, Christian Stockmann, Masataka Asagiri, M Celeste Simon, Alexander Hoffmann, Randall S Johnson
ABSTRACT
Hypoxic response and inflammation both involve the action of the hypoxia-inducible transcription factors HIF-1alpha and HIF-2alpha. Previous studies have revealed that both HIF-alpha proteins are in a number of aspects similarly regulated post-translationally. However, the functional interrelationship of these two isoforms remains largely unclear. The polarization of macrophages controls functionally divergent processes; one of these is nitric oxide (NO) production, which in turn is controlled in part by HIF factors. We show here that the HIF-alpha isoforms can be differentially activated: HIF-1alpha is induced by Th1 cytokines in M1 macrophage polarization, whereas HIF-2alpha is induced by Th2 cytokines during an M2 response. This differential response was most evident in polarized macrophages through HIF-alpha isoform-specific regulation of the inducible NO synthase gene by HIF-1alpha, and the arginase1 gene by HIF-2alpha. In silico modeling predicted that regulation of overall NO availability is due to differential regulation of HIF-1alpha versus HIF-2alpha, acting to, respectively, either increase or suppress NO synthesis. An in vivo model of endotoxin challenge confirmed this; thus, these studies reveal that the two homologous transcription factors, HIF-1alpha and HIF-2alpha, can have physiologically antagonistic functions, but that their antiphase regulation allows them to coordinately regulate NO production in a cytokine-induced and transcription-dependent fashion.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by gel-filtration chromatography
Pricing and availability is not currently available.