Skip to Content
Merck
Z-L-Arg 7-amido-4-methylcoumarin hydrochloride

Z-L-Arg 7-amido-4-methylcoumarin hydrochloride

Z-L-Arg 7-amido-4-methylcoumarin hydrochloride
Synonym(s):
Carbobenzoxy-L-arginine-7-amino-4-methylcoumarin hydrochloride, Nα-Cbz-L-arginine 7-amido-4-methylcoumarin hydrochloride, Z-L-Arginine-4-methyl-7-coumarinylamide hydrochloride, Z-Arg-MCA
Empirical Formula (Hill Notation):
C24H27N5O5 · HCl
CAS Number:
Molecular Weight:
501.96
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing