![Zirconium(IV) acetylacetonate 97%](/deepweb/assets/sigmaaldrich/product/structures/368/550/19a27007-75f6-4145-b623-8a2380188ebf/640/19a27007-75f6-4145-b623-8a2380188ebf.png)
Zirconium(IV) acetylacetonate
![Zirconium(IV) acetylacetonate 97%](/deepweb/assets/sigmaaldrich/product/structures/368/550/19a27007-75f6-4145-b623-8a2380188ebf/640/19a27007-75f6-4145-b623-8a2380188ebf.png)
Synonym(s):
2,4-Pentanedione zirconium(IV) derivative, Tetrakis(2,4-pentanedionato)zirconium(IV), Zirconium(IV) 2,4-pentanedionate, Zirconium(IV) acetylacetonate
Linear Formula:
Zr(C5H7O2)4
CAS Number:
Molecular Weight:
487.66
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing