Skip to Content
Merck
Zanamivir certified reference material, pharmaceutical secondary standard

Zanamivir

Zanamivir certified reference material, pharmaceutical secondary standard
Synonym(s):
4-Guanidino-2,4-dideoxy-2,3-dehydro-N-acetylneuraminic acid, 4-Guanidino-Neu5Ac2en
Empirical Formula (Hill Notation):
C12H20N4O7
CAS Number:
Molecular Weight:
332.31

Products

Product Category
Brand
Color
Feature
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing