Skip to Content
Merck
Xanthan from Xanthomonas campestris

Xanthan from Xanthomonas campestris

Xanthan from Xanthomonas campestris
Synonym(s):
Xanthan gum from Xanthomonas campestris, Gum xanthan
CAS Number:
EC Number:

Products

Product Category
Brand
Biological Source
Color
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing