Skip to Content
Merck
WR-1065 ≥98% (HPLC)

WR-1065

WR-1065 ≥98% (HPLC)
Synonym(s):
2-[(3-Aminopropyl)amino]ethanethiol dihydrochloride, WR-1065 dihydrochloride
Empirical Formula (Hill Notation):
C5H14N2S · 2HCl
CAS Number:
Molecular Weight:
207.16

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing