Skip to Content
Merck
Vanillyl butyl ether ≥96%, FG

Vanillyl butyl ether

Vanillyl butyl ether ≥96%, FG
Synonym(s):
4-(butoxymethyl)-2-methoxy-Phenol
Linear Formula:
4-(HO)C6H3-3-(OCH3)CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
210.27

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing