Valsartan Related Compound C
Synonym(s):
N-(1-Oxopentyl)-N-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-L-valine phenylmethyl ester, (S)-N-Valeryl-N-([2′-(1H-tetrazole-5-yl)-biphenyl-4-yl]methyl)valine benzyl ester
Empirical Formula (Hill Notation):
C31H35N5O3
Molecular Weight:
525.64
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing