Skip to Content
Merck
Valacyclovir Related Compound C certified reference material, pharmaceutical secondary standard

Valacyclovir Related Compound C

Valacyclovir Related Compound C certified reference material, pharmaceutical secondary standard
Synonym(s):
2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-methyl-L-valinate hydrochloride, N-Methyl-L-valine 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester hydrochloride
Empirical Formula (Hill Notation):
C14H22N6O4 · HCl
CAS Number:
Molecular Weight:
374.82

Products

Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1