Skip to Content
Merck
Urea solution suitable for microbiology, 40% in H2O

Urea solution

Urea solution suitable for microbiology, 40% in H2O
Synonym(s):
Carbamide, Carbonyldiamide
Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06

Products




Showing 1-3 of 3
Compare
Product Number
Description
Pricing