![TSKgel® ODS-100V HPLC Column phase C18 (octadecyl), L × I.D. 25 cm × 2 mm, 5 μm particle size](/deepweb/assets/sigmaaldrich/product/images/151/731/6c30e02d-13a2-412f-9335-50d34374bc4d/640/6c30e02d-13a2-412f-9335-50d34374bc4d.jpg)
TSKgel® ODS-100V (5 µm) HPLC Columns
![TSKgel® ODS-100V HPLC Column phase C18 (octadecyl), L × I.D. 25 cm × 2 mm, 5 μm particle size](/deepweb/assets/sigmaaldrich/product/images/151/731/6c30e02d-13a2-412f-9335-50d34374bc4d/640/6c30e02d-13a2-412f-9335-50d34374bc4d.jpg)
Synonym(s):
TSKgel® Reversed Phase HPLC Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2