![Trolamine impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/187/330/10b99f7f-554f-4861-83ad-38673d95103d/640/10b99f7f-554f-4861-83ad-38673d95103d.png)
Trolamine impurity B
![Trolamine impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/187/330/10b99f7f-554f-4861-83ad-38673d95103d/640/10b99f7f-554f-4861-83ad-38673d95103d.png)
Synonym(s):
Diethanolamine, 2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine
Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing