Triticonazole
Synonym(s):
(RS)-(E)-5-(4-Chlorobenzylidene)-2,2-dimethyl-1-(1H-1,2,4-triazol-1-ylmethyl)cyclopentanol
Empirical Formula (Hill Notation):
C17H20ClN3O
CAS Number:
Molecular Weight:
317.81
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2