(Trimethylsilyl)methylamine
Synonym(s):
(Aminomethyl)trimethylsilane
Empirical Formula (Hill Notation):
C4H13NSi
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing