![Triamcinolone acetonide British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/233/972/dd7c0c10-53b6-4107-8bc7-052c20922a60/640/dd7c0c10-53b6-4107-8bc7-052c20922a60.png)
Triamcinolone Acetonide
![Triamcinolone acetonide British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/233/972/dd7c0c10-53b6-4107-8bc7-052c20922a60/640/dd7c0c10-53b6-4107-8bc7-052c20922a60.png)
Synonym(s):
Triamcinolone acetonide, 9α-Fluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide, 9α-Fluoro-16α-hydroxyprednisolone 16α,17α-acetonide
Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing