Travoprost Related Compound A
Synonym(s):
(+)-Fluprostenol solution
Empirical Formula (Hill Notation):
C23H29F3O6
CAS Number:
Molecular Weight:
458.47
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing