Travoprost
Synonym(s):
Travoprost solution
Empirical Formula (Hill Notation):
C26H35F3O6
CAS Number:
Molecular Weight:
500.55
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing