Skip to Content
Merck
trans-4-Hydroxy-L-proline analytical standard

trans-4-Hydroxy-L-proline

trans-4-Hydroxy-L-proline analytical standard
Synonym(s):
(2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, Hyp
Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Reference Material Type
Special Grade
Technique