![Tralomethrin mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/109/374/708a8c09-37a2-404d-a60c-366b7bc43be5/640/708a8c09-37a2-404d-a60c-366b7bc43be5.png)
Tralomethrin
![Tralomethrin mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/109/374/708a8c09-37a2-404d-a60c-366b7bc43be5/640/708a8c09-37a2-404d-a60c-366b7bc43be5.png)
Synonym(s):
(S)-α-Cyano-3-phenoxybenzyl (1R,3S)-2,2-dimethyl-3-[(RS)-1,2,2,2-tetrabromoethyl]cyclopropanecarboxylate
Empirical Formula (Hill Notation):
C22H19Br4NO3
CAS Number:
Molecular Weight:
665.01
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing