![Tralkoxydim certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/287/142/e3eff73c-e953-4f31-b504-095751a79e7b/640/e3eff73c-e953-4f31-b504-095751a79e7b.png)
Tralkoxydim
![Tralkoxydim certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/287/142/e3eff73c-e953-4f31-b504-095751a79e7b/640/e3eff73c-e953-4f31-b504-095751a79e7b.png)
Empirical Formula (Hill Notation):
C20H27NO3
CAS Number:
Molecular Weight:
329.43
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing