Ticlopidine hydrochloride
Synonym(s):
5-(o-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
Empirical Formula (Hill Notation):
C14H14ClNS · HCl
CAS Number:
Molecular Weight:
300.25
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3