
Thioguanine

Synonym(s):
6-Thioguanine, 2-Amino-6-mercaptopurine, 2-Amino-6-purinethiol
Empirical Formula (Hill Notation):
C5H5N5S
CAS Number:
Molecular Weight:
167.19
Products
Brand
Formula Weight
Melting Point (°C)
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing